Experiment data/files
The Applied Biosystems™ Analysis Software can import and analyze experiment files (EDS and SDS) that are generated by various Thermo Fisher Scientific real-time qPCR instruments. Every consumable run on a Thermo Fisher Scientific real-time qPCR instrument requires the creation of one or more experiment files that store the associated data. Each experiment file is a virtual representation of a specific consumable (plate, array, or chip) that contains data for all aspects of the qPCR experiment.
Experiment files contain the following information:
  • Target information and arrangement on the plate
  • Sample information and arrangement on the plate
  • Method parameters for the run
File compatibility
The Applied Biosystems™ Software can import data the following experiment file formats generated by Applied Biosystems™ real-time qPCR instruments:
IMPORTANT! The Applied Biosystems™ Analysis Software can import and analyze data from unsupported versions of the instrument software, but we cannot guarantee the performance of the software or provide support for the analyses.
Real-Time PCR System
Supported software version(s)
File extension
Applied Biosystems™ 7900 HT Fast Real-Time PCR System
v2.4 or later
SDS
Applied Biosystems™ 7500 and 7500 Fast Real-Time PCR System
v1.4.1 or later
v2.0.5 or later
EDS
Applied Biosystems™ StepOne™ and StepOnePlus™ Real-Time PCR System
v2.0.1, v2.1, or later
Applied Biosystems™ ViiA™ 7 Real-Time PCR System
v1.1 or later
Applied Biosystems™ QuantStudio™ 12K Flex Real-Time PCR System
v1.1.1 or later
Applied Biosystems™ QuantStudio™ 1 Real-Time PCR System
v1.0 or later
Applied Biosystems™ QuantStudio™ 3 Real-Time PCR System
v1.0 or later
Applied Biosystems™ QuantStudio™ 5 Real-Time PCR System
Applied Biosystems™ QuantStudio™ 6 Flex Real-Time PCR System
v1.0 or later
Applied Biosystems™ QuantStudio™ 6 Pro Real-Time PCR System
Applied Biosystems™ QuantStudio™ 7 Flex Real-Time PCR System
Applied Biosystems™ QuantStudio™ 7 Pro Real-Time PCR System
7500 and 7900HT System standard curve experiment files
By design, the Applied Biosystems™ Analysis Software reads older 7500 and 7900HT System standard curve experiment (SDS) files with the following characteristics as genotyping experiment files. To configure the files to read as standard curve experiments, rename the detectors that are used by the files so that the names do not contain "FAM" or "VIC".
Property
Value
Experiment type
SC (standard curve)
Instrument type
7500 or 7900
Plate setup
Non-empty wells contain exactly two (2) detectors, where:
Detector 1 configuration:
  • Name: FAM
  • Reporter Dye: FAM™ dye
Detector 2 configuration:
  • Name: VIC
  • Reporter Dye: VIC™ dye