Multicomponent plot
The Multicomponent Plot is a plot of the complete spectral contribution of each dye for the selected well during the PCR run.
-
Toolbar – Contains the following tools for controlling the plot:
– Select individual data points from the plot.
– Allows you to click and manually move the position of the plot.
– Zoom the plot to the selected area.
– Zooms out the plot to show all data points.
-
Target/Sample dropdown list – Selects the data (target- or sample-centric) displayed by the plot.
-
Normalized fluorescence – Displays the normalized fluorescence for all wells throughout the thermal cycling protocol.
When you analyze your own experiment, confirm the following:
-
The passive reference dye fluorescence level should remain relatively constant throughout the PCR process.
-
The reporter dye fluorescence level should display a flat region corresponding to the baseline, followed by a rapid rise in fluorescence as the amplification proceeds.
-
There should not be any spikes, dips, and/or sudden changes in the fluorescent signal.
-
There should not be any amplification in negative control wells.
– Saves the plot as an image (PNG or JPG).
– Allows you to adjust the display options for the plot.