About the quality data summary
The quality summary displays a table of the quality flags supported by the software. For each sample, target, or plate, the table lists the flag frequency and location for any experiment that is added to a project. For each quality flag, the table displays the number of times the flag was triggered by the project data. To examine the data that triggered the flag, click the link in the Name column to view the amplification data for the related target, sample, or plate.
In response to the presence of quality flags, consider the following resolutions: