Add sequences to a project
-
Select the project in which you want to add sequences, then add the sequences to the project using one of the two following methods.
-
Upload a file containing your sequences by either clicking or by dragging and dropping your file in the central pane.
After you click Upload DNA Sequences, navigate to the location where your file is saved, select it, and click Open.
-
Enter manually a DNA sequence by clicking .
The list of your sequences is displayed in the central pane with their names, construct ID, lengths, and statuses.
-
- (Optional) Filter the sequences by clicking Passed or Changes Required to display only the sequences that meet or do not meet the GeneArt™ Strings™ criteria, respectively.
- (Optional) Delete unwanted sequences by selecting their names, then by selecting .
- (Optional) Move sequences from one project to another project:
- Click Explore the problems to proceed to the sequence analysis.