Delete an E1–ClipTip™ Pipette
Deleting an E1–ClipTip™ Pipette from the My Pipette™ Creator removes the link to the pipette being removed. You will no longer be able to access programs on the deleted E1–ClipTip™ Pipette from the My Pipette™ Creator application.
-
In the My
Pipette™ Creator home screen, click an online pipette.
Icons of online pipettes are displayed in color.
-
Click Delete the pipette on the information tab.
You will receive an email confirming the deletion of the pipette.
- To delete the link from the E1–ClipTip™ Pipette, on the E1–ClipTip™ Pipette screen, select .