In the My
Pipette™ Creator Application home screen, click Check for E1-ClipTip Software Updates.
The next screen will show the status of each instrument. If the software is up to date, click OK to return to the My
Pipette™ Creator home screen.
(Optional) View the release notes for the software to see changes in the updated version (see Document and support).
Click the checkbox of the pipette to be updated, then click OK.
(Optional) Click Edit My Selection to change the pipette being updated.