My Pipette Creator Application Help
About My Pipette™ Creator
The My Pipette™ Creator Application is an application for managing pipetting programs and software for E1–ClipTip™ Pipettes.
Note:
At any time, click My Pipette Creator Home to return to the My Pipette™ Creator home page.
Function
Topic
1
Protocol Library tab: Search and download available kit protocols.
Select kit‑specific protocols for the E1–ClipTip Pipette
2
My Pipettes tab: Shows the pipettes connected to your Thermo Fisher Cloud account.
Manage instruments and software
3
Add a new E1–ClipTip™ button: Connect a new E1–ClipTip™ Pipette with your Thermo Fisher Cloud account.
Add an E1–ClipTip Pipette
4
Create Program button: Create or edit programs.
Create or edit programs
5
Transfer Programs button: Move programs between the Thermo Fisher Cloud (My Programs) and a connected E1–ClipTip™ Pipette.
Transfer programs
6
Programs list: Shows the programs under the selected tab.
See callout 2, 9, or 10.
7
Recent Programs tab: Shows the most recently used programs. Click a program to edit.
Create or edit programs
8
Thermo Fisher Cloud icons
General icons for the Thermo Fisher Cloud are in the black bar on the right of the main My Pipette™ Creator home page. See the help system for the Thermo Fisher Cloud.
9
My Programs tab: Shows programs saved to your Thermo Fisher Cloud account. Click a program to edit.
Working with programs
10
Shared Programs tab: Shows the programs being shared by other users with you.
Working with programs