Matrix steps
The following steps are available when creating or editing a Matrix pipetting program.
Step |
Use the step to: |
---|---|
Fill |
Fill the tip with liquid. |
Dispense |
Dispense liquid. |
Multi |
Repeatedly dispense liquid. |
Purge |
Empty the tip. |
Mix |
Mix by pipette using a selected volume. |
Air gap |
Select a volume of air to separate 2 liquids in the pipette. |
Auto |
Dispense the same volume repeatedly over a defined time interval. |
Timer |
Select the time before the next step can be performed. |
Note text |
Text that describes the step. The text is displayed on the screen during the step. |
Tip eject |
Ejects pipette tips.[1] |
Loop |
Repeats a series of steps. |