Example workflow used with the SeqScreener
Gene Edit Confirmation App
From experiment set up to result
Set up experiments on Thermo Fisher™Connect
OR
Set up experiments on your capillary electrophoresis (CE)
Sanger sequencing instrument
Use the Plate Manager software available through Thermo Fisher™Connect or set up experiments directly on
your CE instrument. Refer to the software Help or the instrument User Guide for guidance.
Run samples on a CE Sanger sequencing instrument
For instructions on running samples on a CE instrument, refer to the user guide
specific to your instrument.
Set up your project and import files for analysis in SeqScreener
Gene Edit Confirmation App (see Start a new project)
Use the Setup tab to
start a new project, upload sample and control files, enter sample information, and
launch an analysis.
View the summary of editing efficiency results in the
Results tab and use
the interactive visualization tools in the
Details tab to
evaluate the editing efficiency quality for each sample in a project.