Start a new project
Use the Setup tab to start a new project, upload sample and control files, enter the required sample information, and proceed to analysis. When you open the SeqScreener Gene Edit Confirmation App from Thermo Fisher™ Connect, the Setup tab opens by default. If you are viewing results from the Results tab or the Details tab and would like to start a new project or add more samples to an existing project, you can go back to project setup by clicking the Setup tab.
Note: When you start a new project, the project name is automatically assigned based on the naming convention designated during application set up (out-of-box) or in Settings. For more information, including how to customize your project naming convention, see Customize project and application settings.