View detailed results for a project
-
Navigate to the results details using one of
the following approaches.
In the Results tab, click within the row of a sample of interest to open the Details tab with the desired sample pre-selected in the Overview screen.
By default, the Overview screen opens. If you clicked within the sample row in the Results tab, the corresponding well is automatically selected in the Plate overview.
- Use one of the following approaches to navigate between sample results.
- Click Overview, Indel Frequency Chart, Donor to Control Alignment, or Sequencing Traces to visualize results for the sample selected in step 2.
Results details are organized in the following screens.
Screen |
Description |
---|---|
View results in a plate layout format, which provides a visual overview of editing quality for each sample in a project. For more information, see Results details: Overview (plate view). |
|
View DNA sequence distributions and compare sequences for a sample. For more information, see Results details: Indel frequency chart. |
|
View the alignment of donor DNA to control DNA sequence for a sample. For more information, see Results details: Donor to control alignment. |
|
View sequencing traces of an edited sample and its control side by side. For more information, see Results details: Sequencing traces. |
To export results, see Export results details.