Sizing Analysis Module Peak Scanner Software Help
About the Sizing Analysis Module Peak Scanner™ Software
Sizing Analysis Module Peak Scanner™ Software is a DNA sizing module available on Thermo Fisher Cloud.
Use this software to perform DNA fragment analysis, separate a mixture of DNA fragments according to their sizes, provide a profile of the separation, and precisely calculate the sizes of the fragments.
The software allows you to view, edit, analyze, print, and export fragment analysis data generated using Applied Biosystems™ genetic analyzers.