JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Compatible instruments Capillary Electrophoresis System Supported software version(s) File extension Applied Biosystems™ SeqStudio™ Genetic Analyzer (3200) v1.0 or later FSA Applied Biosystems™ 3500/3500xL Genetic Analyzer Applied Biosystems™ 3130/3130xl Genetic Analyzer Applied Biosystems™ 3730/3730xl DNA Analyzer Parent topic: About the Sizing Analysis Module Peak Scanner™ Software