Experiment data/files
The Applied Biosystems™ Analysis Software can import and analyze experiment files (EDS and SDS) that are generated by various Thermo Fisher Scientific real-time qPCR instruments. Every consumable run on a Thermo Fisher Scientific real-time qPCR instrument requires the creation of one or more experiment files that store the associated data. Each experiment file is a virtual representation of a specific consumable (plate, array, or chip) that contains data for all aspects of the qPCR experiment.
Experiment files contain the following information:
  • Target information and arrangement on the plate
  • Sample information and arrangement on the plate
  • Method parameters for the run
File compatibility
The Applied Biosystems™ Software can import data the following experiment file formats generated by Applied Biosystems™ real-time qPCR instruments:
IMPORTANT! The Applied Biosystems™ Analysis Software can import and analyze data from unsupported versions of the instrument software, but we cannot guarantee the performance of the software or provide support for the analyses.
Real-Time PCR System
Supported software version(s)
File extension
Applied Biosystems™ 7900 HT Fast Real-Time PCR System
v2.4 or later
SDS
Applied Biosystems™ 7500 and 7500 Fast Real-Time PCR System
v1.4.1 or later
v2.0.5 or later
EDS
Applied Biosystems™ StepOne™ and StepOnePlus™ Real-Time PCR System
v2.0.1, v2.1, or later
Applied Biosystems™ ViiA™ 7 Real-Time PCR System
v1.1 or later
Applied Biosystems™ QuantStudio™ 12K Flex Real-Time PCR System
v1.1.1 or later
Applied Biosystems™ QuantStudio™ 1 Real-Time PCR System
v1.0 or later
Applied Biosystems™ QuantStudio™ 3 Real-Time PCR System
v1.0 or later
Applied Biosystems™ QuantStudio™ 5 Real-Time PCR System
Applied Biosystems™ QuantStudio™ 6 Flex Real-Time PCR System
v1.0 or later
Applied Biosystems™ QuantStudio™ 6 Pro Real-Time PCR System
Applied Biosystems™ QuantStudio™ 7 Flex Real-Time PCR System
Applied Biosystems™ QuantStudio™ 7 Pro Real-Time PCR System