Manage your projects and experiment data
Use the Data Manager screen to add and remove experiments to and from your project. The screen displays all experiments associated with the current project. You can also use the Data Manager to upload new .eds and .sds files or view the details of individual experiments already added to the project.
To view the Data Manager screen, click in the toolbar on the left side of any other screen.
Tips for using the Data Manager
-
Click the column headings of the Data Manager table to sort the contents. The direction of the sort is indicated by the presence of an arrow ( or ) in the header.
-
The Instrument column indicates the real-time PCR instrument used to generate the data.
-
The Size column indicates the data footprint of each object in kilobytes.
-
The Run Date column indicates the date and time that the experiment data was collected.