Create a new program
- In the My Pipette™ Creator Home screen, select .
- Enter a program name.
- Select your pipette model.
-
Select a program type from the dropdown list.
Program type
Description
Matrix
Preset pipetting functions
-
(Optional) Enter keywords in the Tags field.
Tags are keywords that you can use to search for your program.
- (Optional) Enter comments in the Description field.
-
Click Continue.
The programming screen appears.