Edit a Preset pipetting function parameter
Before editing a parameter of a Preset pipetting function, you must create a new program or select an program to edit. See one of the following topics for instructions.
-
In the programming screen, click in a field to change a parameter.
See Preset pipetting functions or the E1–ClipTip™ Pipette user guide for additional information.
- Click Save.