To omit the data from one or more wells that you do not want included in the analysis:
IMPORTANT! You cannot omit all wells that belong to a reference sample, that belong to a biological group, or that serve as the endogenous control for the project.
Note: To restore an omitted well, select the well from a plot or table, then select .