In comparative CT (∆∆CT) experiments or gene expression studies, the sample used as the basis of comparison for relative quantification results. Also called the calibrator.
For a gene expression project using biological groups, a biological group is used as the reference rather than a sample.